siRNA Details
Gene Symbol: SMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017411.3 | NP_059107.1 | 4 | 628 | ||
NM_022875.2 | NP_075013.1 | 4 | 628 | |||
NM_022876.2 | NP_075014.1 | 4 | 628 | |||
NM_022877.2 | NP_075015.1 | 4 | 628 | |||
XM_011543599.1 | XP_011541901.1 | 4 | 507 | |||
XM_017009787.1 | XP_016865276.1 | 4 | 507 | |||
GenBank mRNA | AA029652.1 | - | 1 | 18 | ||
CN272420.1 | - | 2 | 331 | |||
JQ690865.1 | - | 2 | 217 |
Gene Symbol: SMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000344.3 | NP_000335.1 | 4 | 628 | ||
NM_001297715.1 | NP_001284644.1 | 4 | 628 | |||
NM_022874.2 | NP_075012.1 | 4 | 628 | |||
XM_011543596.1 | XP_011541898.1 | 4 | 507 | |||
XM_017009786.1 | XP_016865275.1 | 4 | 507 | |||
GenBank mRNA | BC062723.1 | - | 4 | 484 | 4250429 | |
JQ657802.1 | - | 2 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
