siRNA Details
Gene Symbol: SMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017411.3 | NP_059107.1 | 5 | 651 | ||
NM_022875.2 | NP_075013.1 | 5 | 651 | |||
NM_022876.2 | NP_075014.1 | 5 | 651 | |||
NM_022877.2 | NP_075015.1 | 5 | 651 | |||
XM_011543599.1 | XP_011541901.1 | 5 | 530 | |||
XM_011543600.2 | XP_011541902.1 | 4 | 335 | |||
XM_011543601.1 | XP_011541903.1 | 4 | 335 | |||
XM_011543602.2 | XP_011541904.1 | 4 | 347 | |||
XM_011543603.2 | XP_011541905.1 | 4 | 347 | |||
XM_017009787.1 | XP_016865276.1 | 5 | 530 | |||
GenBank mRNA | AA029652.1 | - | 2 | 41 | ||
AK289669.1 | - | 5 | 534 | |||
BC000908.2 | - | 5 | 520 | 2985046 | ||
BC015308.1 | - | 5 | 518 | 4427000 | ||
BC070242.1 | - | 5 | 497 | 4673318 | ||
CN272420.1 | - | 3 | 354 | |||
JQ690861.1 | - | 2 | 39 | |||
JQ690862.1 | - | 2 | 39 | |||
JQ690863.1 | - | 2 | 39 | |||
JQ690865.1 | - | 3 | 240 | |||
JQ690866.1 | - | 2 | 39 | |||
JQ732167.1 | - | 3 | 240 | |||
U21914.1 | - | 5 | 485 |
Gene Symbol: SMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000344.3 | NP_000335.1 | 5 | 651 | ||
NM_001297715.1 | NP_001284644.1 | 5 | 651 | |||
NM_022874.2 | NP_075012.1 | 5 | 651 | |||
XM_011543596.1 | XP_011541898.1 | 5 | 530 | |||
XM_011543597.1 | XP_011541899.1 | 4 | 335 | |||
XM_011543598.2 | XP_011541900.1 | 4 | 346 | |||
XM_017009786.1 | XP_016865275.1 | 5 | 530 | |||
GenBank mRNA | AK298203.1 | - | 4 | 335 | ||
BC062723.1 | - | 5 | 507 | 4250429 | ||
JQ657798.1 | - | 3 | 240 | |||
JQ657799.1 | - | 3 | 240 | |||
JQ657800.1 | - | 2 | 39 | |||
JQ657801.1 | - | 3 | 240 | |||
JQ657802.1 | - | 3 | 240 | |||
JQ657803.1 | - | 2 | 39 | |||
JQ690867.1 | - | 2 | 39 | |||
JQ690868.1 | - | 2 | 39 | |||
JQ732166.1 | - | 3 | 240 | |||
U18423.1 | - | 5 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
