siRNA Details
Gene Symbol: SMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017411.3 | NP_059107.1 | 2, 3 | 319 | ||
NM_022875.2 | NP_075013.1 | 2, 3 | 319 | |||
NM_022876.2 | NP_075014.1 | 2, 3 | 319 | |||
NM_022877.2 | NP_075015.1 | 2, 3 | 319 | |||
XM_011543599.1 | XP_011541901.1 | 2, 3 | 198 | |||
XM_011543600.2 | XP_011541902.1 | 2, 3 | 204 | |||
XM_011543601.1 | XP_011541903.1 | 2, 3 | 204 | |||
XM_011543602.2 | XP_011541904.1 | 2, 3 | 216 | |||
XM_011543603.2 | XP_011541905.1 | 2, 3 | 216 | |||
XM_017009787.1 | XP_016865276.1 | 2, 3 | 198 | |||
GenBank mRNA | AK289669.1 | - | 2, 3 | 202 | ||
BC000908.2 | - | 2, 3 | 188 | 2985046 | ||
BC015308.1 | - | 2, 3 | 186 | 4427000 | ||
BC070242.1 | - | 2, 3 | 165 | 4673318 | ||
U21914.1 | - | 2, 3 | 153 |
Gene Symbol: SMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000344.3 | NP_000335.1 | 2, 3 | 319 | ||
NM_001297715.1 | NP_001284644.1 | 2, 3 | 319 | |||
NM_022874.2 | NP_075012.1 | 2, 3 | 319 | |||
XM_011543596.1 | XP_011541898.1 | 2, 3 | 198 | |||
XM_011543597.1 | XP_011541899.1 | 2, 3 | 204 | |||
XM_011543598.2 | XP_011541900.1 | 2, 3 | 215 | |||
XM_017009786.1 | XP_016865275.1 | 2, 3 | 198 | |||
GenBank mRNA | AK298203.1 | - | 2, 3 | 204 | ||
BC062723.1 | - | 2, 3 | 175 | 4250429 | ||
U18423.1 | - | 2, 3 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
