siRNA Details
Gene Symbol: SMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000344.3 | NP_000335.1 | 8 | 1023 | ||
NM_022874.2 | NP_075012.1 | 7 | 927 | |||
XM_011543597.1 | XP_011541899.1 | 7 | 707 | |||
XM_011543598.2 | XP_011541900.1 | 6 | 622 | |||
GenBank mRNA | AK298203.1 | - | 7 | 707 | ||
BC062723.1 | - | 8 | 879 | 4250429 | ||
JQ657798.1 | - | 5 | 516 | |||
JQ657800.1 | - | 5 | 411 | |||
JQ657802.1 | - | 5 | 516 | |||
JQ657803.1 | - | 4 | 315 | |||
JQ732166.1 | - | 4 | 405 | |||
JQ745297.1 | - | 4 | 258 | |||
U18423.1 | - | 8 | 893 |
Gene Symbol: SMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017411.3 | NP_059107.1 | 8 | 1023 | ||
NM_022876.2 | NP_075014.1 | 7 | 927 | |||
XM_011543600.2 | XP_011541902.1 | 7 | 707 | |||
XM_011543602.2 | XP_011541904.1 | 6 | 623 | |||
GenBank mRNA | BC015308.1 | - | 8 | 890 | 4427000 | |
CN272420.1 | - | 6 | 726 | |||
JQ690861.1 | - | 5 | 411 | |||
JQ690866.1 | - | 4 | 315 | |||
JQ732167.1 | - | 4 | 405 | |||
U21914.1 | - | 8 | 857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
