siRNA Details
Gene Symbol: FAM156B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099684.2 | NP_001093154.1 | 1 | 51 | ||
NM_001321178.1 | NP_001308107.1 | 1 | 1814 | |||
NM_001321180.1 | NP_001308109.1 | 2 | 211 | |||
NM_001321181.1 | NP_001308110.1 | 2 | 183 | |||
NM_001321182.1 | NP_001308111.1 | 2 | 165 | |||
XM_011530817.1 | XP_011529119.1 | 2 | 165 | |||
GenBank mRNA | BX649013.1 | - | 1 | 1777 |
Gene Symbol: FAM156A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242489.1 | NP_001229418.1 | 2 | 218 | ||
NM_001242490.1 | NP_001229419.1 | 2 | 109 | |||
NM_001242491.1 | NP_001229420.1 | 2 | 165 | |||
NM_014138.4 | NP_054857.2 | 3 | 260 | |||
XM_006724586.3 | XP_006724649.1 | 2 | 1357 | |||
XM_011530781.1 | XP_011529083.1 | 2 | 165 | |||
XM_017029457.1 | XP_016884946.1 | 3 | 2116 | |||
GenBank mRNA | AF370413.1 | - | 1 | 274 | ||
AK057692.1 | - | 2 | 148 | |||
AK096162.1 | - | 1 | 560 | |||
BU192418.1 | - | 2 | 90 | |||
DA572135.1 | - | 3 | 260 | |||
DA849151.1 | - | 2 | 139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
