siRNA Details
Gene Symbol: FAM156B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099684.2 | NP_001093154.1 | 3 | 794 | ||
NM_001321178.1 | NP_001308107.1 | 3 | 2560 | |||
NM_001321179.1 | NP_001308108.1 | 4 | 1819 | |||
NM_001321180.1 | NP_001308109.1 | 4 | 957 | |||
NM_001321181.1 | NP_001308110.1 | 4 | 929 | |||
NM_001321182.1 | NP_001308111.1 | 3 | 818 | |||
NM_001321183.1 | NP_001308112.1 | 4 | 638 | |||
NM_001321186.1 | NP_001308115.1 | 4 | 520 | |||
NM_001321187.1 | NP_001308116.1 | 4 | 502 | |||
NM_001321188.1 | NP_001308117.1 | 4 | 446 | |||
XM_011530817.1 | XP_011529119.1 | 4 | 911 | |||
XM_011530821.2 | XP_011529123.1 | 3 | 419 | |||
GenBank mRNA | AK091598.1 | - | 4 | 1715 | ||
BX649013.1 | - | 3 | 2523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
