siRNA Details
Gene Symbol: ZNF737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159293.1 | NP_001152765.1 | 4 | 1587 | ||
XM_005259697.3 | XP_005259754.1 | 4 | 1576 | |||
XM_006722597.3 | XP_006722660.1 | 4 | 1576 | |||
XM_011527598.2 | XP_011525900.1 | 4 | 1576 | |||
XM_011527599.2 | XP_011525901.1 | 4 | 1576 | |||
XM_017026116.1 | XP_016881605.1 | 5 | 1674 | |||
XM_017026117.1 | XP_016881606.1 | 3 | 1461 |
Gene Symbol: ZNF626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076675.2 | NP_001070143.1 | 4 | 1976 | ||
GenBank mRNA | AK096221.1 | - | 3 | 1535 | ||
BC150651.1 | - | 5 | 1720 | 9057022 | ||
BX648764.1 | - | 3 | 1562 | |||
CR749233.1 | - | 5 | 1761 |
Gene Symbol: ZNF738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027130.1 | 5 | 8702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
