siRNA Details
Gene Symbol: LYRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164841.2 | NP_001158313.1 | 4 | 820 | ||
NM_020408.5 | NP_065141.3 | 3 | 695 | |||
NR_104417.1 | 4 | 1042 | ||||
NR_104418.1 | 2 | 574 | ||||
GenBank mRNA | AF088060.1 | - | 3 | 475 | ||
AF170070.1 | - | 3 | 491 | |||
AF258559.1 | - | 1 | 14 | |||
AF285118.1 | - | 3 | 491 | |||
AK291158.1 | - | 3 | 487 | |||
AK309458.1 | - | 3 | 741 | |||
BC009552.2 | - | 3 | 546 | 3898393 | ||
BC067762.1 | - | 2 | 366 | |||
BC105601.1 | - | 2 | 310 | |||
BI758258.1 | - | 3 | 685 | |||
BX353584.2 | - | 4 | 599 | |||
BX446965.2 | - | 1 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
