siRNA Details
Gene Symbol: TMEM203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053045.1 | NP_444273.1 | 1 | 1001 | ||
GenBank mRNA | AF529372.1 | - | 1 | 42 | ||
AX775751.1 | - | 1 | 1010 | |||
BC009283.2 | - | 1 | 1001 | 4298098 | ||
BC009461.1 | - | 1 | 1001 | 3538818 | ||
BC067786.1 | - | 1 | 1013 | 5275460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
