siRNA Details
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127395.2 | NP_001120867.1 | 4 | 2300 | ||
NM_145280.5 | NP_660323.3 | 4 | 2161 |
Gene Symbol: FKSG48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336875.1 | - | 1 | 253 |
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 178 | ||
AF336879.1 | - | 1 | 202 | |||
AF336883.1 | - | 1 | 187 | |||
AF336884.1 | - | 1 | 247 | |||
AF336886.1 | - | 1 | 175 | |||
AF336887.1 | - | 1 | 232 | |||
AF338193.1 | - | 1 | 238 | |||
AF338194.1 | - | 1 | 241 | |||
AF338196.1 | - | 1 | 250 | |||
AF338198.1 | - | 1 | 238 | |||
AF338199.1 | - | 1 | 205 | |||
AF469044.1 | - | 1 | 187 |
Gene Symbol: LOC100506290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336880.1 | - | 1 | 259 |
Gene Symbol: LOC402469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336877.1 | - | 1 | 250 | ||
AF336881.1 | - | 1 | 250 | |||
AF338191.1 | - | 1 | 235 |
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024077.1 | - | 4 | 1510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
