siRNA Details
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 224 | ||
AF336879.1 | - | 1 | 248 | |||
AF336883.1 | - | 1 | 233 | |||
AF336884.1 | - | 1 | 293 | |||
AF336886.1 | - | 1 | 221 | |||
AF336887.1 | - | 1 | 278 | |||
AF338190.1 | - | 1 | 266 | |||
AF338193.1 | - | 1 | 284 | |||
AF338194.1 | - | 1 | 287 | |||
AF338195.1 | - | 1 | 284 | |||
AF338198.1 | - | 1 | 284 | |||
AF338199.1 | - | 1 | 251 | |||
AF469044.1 | - | 1 | 233 |
Gene Symbol: FKSG48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336875.1 | - | 1 | 299 |
Gene Symbol: LOC100506290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336880.1 | - | 1 | 305 |
Gene Symbol: LOC402469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336877.1 | - | 1 | 296 | ||
AF336881.1 | - | 1 | 296 | |||
AF338191.1 | - | 1 | 281 |
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127395.2 | NP_001120867.1 | 4 | 2346 | ||
NM_145280.5 | NP_660323.3 | 4 | 2207 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126330.1 | - | 9 | 2589 |
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024077.1 | - | 4 | 1556 |
Gene Symbol: SEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209437.1 | - | 1 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
