siRNA Details
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 76 | ||
AF336879.1 | - | 1 | 100 | |||
AF336882.1 | - | 1 | 136 | |||
AF336883.1 | - | 1 | 85 | |||
AF336884.1 | - | 1 | 145 | |||
AF336886.1 | - | 1 | 73 | |||
AF336887.1 | - | 1 | 130 | |||
AF338190.1 | - | 1 | 118 | |||
AF338193.1 | - | 1 | 136 | |||
AF338194.1 | - | 1 | 139 | |||
AF338195.1 | - | 1 | 136 | |||
AF338196.1 | - | 1 | 148 | |||
AF338198.1 | - | 1 | 136 | |||
AF338199.1 | - | 1 | 103 | |||
AF469044.1 | - | 1 | 85 |
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 3776 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 2842 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 3643 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 3330 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 3905 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 4169 | |||
NM_003879.5 | NP_003870.4 | 10 | 3748 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 4643 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 3612 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 3691 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 4578 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 6595 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 4451 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 3789 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 3414 |
Gene Symbol: HELLPAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JX088243.1 | - | 1 | 53284 |
Gene Symbol: LOC100506290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336880.1 | - | 1 | 157 |
Gene Symbol: LOC402469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336877.1 | - | 1 | 148 | ||
AF336881.1 | - | 1 | 148 | |||
AF338191.1 | - | 1 | 133 |
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127395.2 | NP_001120867.1 | 4 | 2198 | ||
NM_145280.5 | NP_660323.3 | 4 | 2059 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199289.2 | NP_954983.1 | 22 | 2782 | ||
GenBank mRNA | AK126330.1 | - | 9 | 2441 | ||
BC063885.1 | - | 22 | 2768 | 30383658 |
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024077.1 | - | 4 | 1408 |
Gene Symbol: SPICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832327.2 | - | 12 | 2923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
