siRNA Details
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 272 | ||
AF336879.1 | - | 1 | 296 | |||
AF336882.1 | - | 1 | 332 | |||
AF336883.1 | - | 1 | 281 | |||
AF336886.1 | - | 1 | 269 | |||
AF336887.1 | - | 1 | 326 | |||
AF338190.1 | - | 1 | 314 | |||
AF338193.1 | - | 1 | 332 | |||
AF338194.1 | - | 1 | 335 | |||
AF338195.1 | - | 1 | 332 | |||
AF338196.1 | - | 1 | 344 | |||
AF338198.1 | - | 1 | 332 | |||
AF338199.1 | - | 1 | 299 | |||
AF469044.1 | - | 1 | 281 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126330.1 | - | 9 | 2637 |
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024077.1 | - | 4 | 1604 |
Gene Symbol: RRM1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | H70418.1 | - | 1 | 82 |
Gene Symbol: SEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209437.1 | - | 1 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
