siRNA Details
Gene Symbol: ANKDD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543618.2 | XP_011541920.1 | 1 | 24 | ||
XM_011543623.2 | XP_011541925.1 | 1 | 24 | |||
XM_017009814.1 | XP_016865303.1 | 1 | 24 | |||
XM_017009815.1 | XP_016865304.1 | 1 | 24 | |||
XM_017009816.1 | XP_016865305.1 | 1 | 24 | |||
XM_017009817.1 | XP_016865306.1 | 1 | 24 |
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 324 | ||
AF336879.1 | - | 1 | 348 | |||
AF336882.1 | - | 1 | 384 | |||
AF336883.1 | - | 1 | 333 | |||
AF336884.1 | - | 1 | 393 | |||
AF336886.1 | - | 1 | 321 | |||
AF338190.1 | - | 1 | 366 | |||
AF338195.1 | - | 1 | 384 | |||
AF338196.1 | - | 1 | 396 | |||
AF338198.1 | - | 1 | 384 | |||
AF469044.1 | - | 1 | 333 |
Gene Symbol: LOC100506290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336880.1 | - | 1 | 405 |
Gene Symbol: LOC402469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336877.1 | - | 1 | 396 | ||
AF336881.1 | - | 1 | 396 | |||
AF338191.1 | - | 1 | 381 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126330.1 | - | 9 | 2689 |
Gene Symbol: SEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209437.1 | - | 1 | 537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
