siRNA Details
Gene Symbol: WDR70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018034.2 | NP_060504.1 | 3 | 186 | ||
XM_005248318.3 | XP_005248375.1 | 3 | 207 | |||
XM_017009604.1 | XP_016865093.1 | 3 | 216 | |||
XM_017009605.1 | XP_016865094.1 | 3 | 186 | |||
GenBank mRNA | AK001095.1 | - | 3 | 128 | ||
AK291179.1 | - | 3 | 208 | |||
AK310901.1 | - | 3 | 211 | |||
AL512685.1 | - | 3 | 188 | |||
BC009648.1 | - | 3 | 186 | 3454326 | ||
BC025315.1 | - | 3 | 164 | 5458850 | ||
BX537655.1 | - | 2 | 143 |
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027026.1 | 3 | 208 | |||
NR_027027.1 | 3 | 208 | ||||
NR_027028.2 | 3 | 208 | ||||
GenBank mRNA | AK095656.1 | - | 3 | 132 | ||
AK290693.1 | - | 3 | 175 | |||
BC094818.1 | - | 3 | 147 | |||
BC147000.1 | - | 3 | 162 | |||
DA358891.1 | - | 3 | 208 |
Gene Symbol: LINC00680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125727.1 | 3 | 236 | |||
NR_125728.1 | 3 | 236 | ||||
NR_125729.1 | 3 | 236 | ||||
NR_132994.1 | 3 | 236 | ||||
NR_132995.1 | 3 | 236 | ||||
NR_132996.1 | 2 | 163 | ||||
GenBank mRNA | AL541195.3 | - | 3 | 169 | ||
BC011838.2 | - | 3 | 170 | |||
BC017330.2 | - | 3 | 164 | |||
BC030061.1 | - | 3 | 164 | |||
BC065547.1 | - | 3 | 194 | |||
BE885753.1 | - | 2 | 79 | |||
BI756196.1 | - | 3 | 170 | |||
DA613065.1 | - | 3 | 216 |
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 3 | 236 | |||
NR_132997.1 | 3 | 236 | ||||
NR_132998.1 | 2 | 163 | ||||
GenBank mRNA | BC144584.1 | - | 3 | 172 | ||
BC144585.1 | - | 2 | 99 | |||
BM553931.1 | - | 3 | 236 | |||
DA269273.1 | - | 3 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
