siRNA Details
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027026.1 | 3 | 231 | |||
NR_027027.1 | 3 | 231 | ||||
NR_027028.2 | 3 | 231 | ||||
GenBank mRNA | AK095656.1 | - | 3 | 155 | ||
AK290693.1 | - | 3 | 198 | |||
BC094818.1 | - | 3 | 170 | |||
BC147000.1 | - | 3 | 185 | |||
DA358891.1 | - | 3 | 231 |
Gene Symbol: LINC00680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125727.1 | 3 | 259 | |||
NR_125728.1 | 3 | 259 | ||||
NR_125729.1 | 3 | 259 | ||||
NR_132994.1 | 3 | 259 | ||||
NR_132995.1 | 3 | 259 | ||||
NR_132996.1 | 2 | 186 | ||||
GenBank mRNA | AL541195.3 | - | 3 | 192 | ||
BC011838.2 | - | 3 | 193 | |||
BC017330.2 | - | 3 | 187 | |||
BC030061.1 | - | 3 | 187 | |||
BC065547.1 | - | 3 | 217 | |||
BE885753.1 | - | 2 | 102 | |||
BI756196.1 | - | 3 | 193 | |||
DA613065.1 | - | 3 | 239 |
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 3 | 259 | |||
NR_132997.1 | 3 | 259 | ||||
NR_132998.1 | 2 | 186 | ||||
GenBank mRNA | BC144584.1 | - | 3 | 195 | ||
BC144585.1 | - | 2 | 122 | |||
BM553931.1 | - | 3 | 259 | |||
DA269273.1 | - | 3 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
