siRNA Details
Gene Symbol: PSG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130167.1 | NP_001123639.1 | 3 | 793 | ||
NM_001130168.1 | NP_001123640.1 | 2 | 427 | |||
NM_182707.2 | NP_874366.1 | 3 | 793 | |||
XM_011526986.1 | XP_011525288.1 | 3 | 806 | |||
XM_011526987.2 | XP_011525289.1 | 2 | 585 | |||
XM_011526988.2 | XP_011525290.1 | 3 | 776 | |||
GenBank mRNA | AK300230.1 | - | 2 | 426 | ||
AL545004.3 | - | 2 | 410 | |||
BC137500.1 | - | 3 | 793 | 9021500 | ||
BC142628.1 | - | 3 | 781 | 40147510 | ||
CR749812.1 | - | 3 | 792 | |||
M30629.1 | - | 2 | 557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
