siRNA Details
Gene Symbol: PSG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130167.1 | NP_001123639.1 | 2 | 361 | ||
NM_182707.2 | NP_874366.1 | 2 | 361 | |||
XM_011526986.1 | XP_011525288.1 | 2 | 374 | |||
GenBank mRNA | BC137500.1 | - | 2 | 361 | 9021500 | |
BC142628.1 | - | 2 | 349 | 40147510 | ||
CR749812.1 | - | 2 | 360 |
Gene Symbol: PSG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002785.2 | NP_002776.3 | 2 | 370 | ||
GenBank mRNA | BF084984.1 | - | 1 | 182 | ||
BG619770.1 | - | 2 | 371 | |||
M69245.1 | - | 2 | 332 |
Gene Symbol: PSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031246.3 | NP_112536.2 | 2 | 435 | ||
GenBank mRNA | BC022316.1 | - | 2 | 362 | 4731250 | |
M20882.1 | - | 2 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
