siRNA Details
Gene Symbol: PSG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130167.1 | NP_001123639.1 | 2 | 389 | ||
NM_182707.2 | NP_874366.1 | 2 | 389 | |||
XM_011526986.1 | XP_011525288.1 | 2 | 402 | |||
GenBank mRNA | BC137500.1 | - | 2 | 389 | 9021500 | |
BC142628.1 | - | 2 | 377 | 40147510 | ||
CR749812.1 | - | 2 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
