siRNA Details
Gene Symbol: PSG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031850.3 | NP_001027020.1 | 5 | 1151 | ||
NM_002782.4 | NP_002773.1 | 5 | 1151 | |||
GenBank mRNA | BC020652.1 | - | 5 | 1118 | 4770562 | |
M31125.1 | - | 5 | 1064 | |||
M33666.1 | - | 5 | 1071 |
Gene Symbol: PSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184825.1 | NP_001171754.1 | 5 | 1160 | ||
NM_001184826.1 | NP_001171755.1 | 5 | 1160 | |||
NM_001297773.1 | NP_001284702.1 | 5 | 1108 | |||
NM_006905.2 | NP_008836.2 | 5 | 1160 | |||
XM_005259065.3 | XP_005259122.1 | 4 | 880 | |||
XM_011527125.1 | XP_011525427.1 | 4 | 880 | |||
XM_017026994.1 | XP_016882483.1 | 4 | 880 | |||
GenBank mRNA | AK056754.1 | - | 2 | 1925 | ||
AK300152.1 | - | 5 | 823 | |||
AK300206.1 | - | 5 | 769 | |||
AK311491.1 | - | 5 | 1061 | |||
BC022338.1 | - | 5 | 1160 | 4732357 | ||
BC058285.1 | - | 5 | 1084 | 4770394 | ||
CR450329.1 | - | 5 | 1022 | |||
J04539.1 | - | 5 | 1082 | |||
M17908.1 | - | 5 | 1088 | |||
M20879.1 | - | 5 | 1108 | |||
M20881.1 | - | 5 | 1139 | |||
M21822.1 | - | 5 | 1095 | |||
M25385.1 | - | 5 | 1058 | |||
M33663.1 | - | 5 | 1085 | |||
M33664.1 | - | 5 | 1022 | |||
M34715.1 | - | 5 | 1183 | |||
M37397.1 | - | 5 | 1112 |
Gene Symbol: PSG10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026824.1 | 4 | 842 | |||
GenBank mRNA | AK309876.1 | - | 4 | 842 |
Gene Symbol: PSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021016.3 | NP_066296.2 | 5 | 1154 | ||
XM_011527126.1 | XP_011525428.1 | 4 | 875 | |||
GenBank mRNA | BC005924.1 | - | 5 | 1119 | 4043628 | |
M23575.1 | - | 5 | 1253 | |||
M27907.1 | - | 2 | 238 | |||
M34420.1 | - | 5 | 1103 | |||
M37399.1 | - | 5 | 1108 | |||
M93061.1 | - | 5 | 1080 | |||
S59493.1 | - | 5 | 1080 |
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276495.1 | NP_001263424.1 | 4 | 981 | ||
NM_001316339.1 | NP_001303268.1 | 4 | 894 | |||
NM_002780.4 | NP_002771.2 | 5 | 1260 | |||
NM_213633.2 | NP_998798.1 | 4 | 981 | |||
XM_017026995.1 | XP_016882484.1 | 6 | 1323 | |||
XM_017026996.1 | XP_016882485.1 | 6 | 1322 | |||
XM_017026997.1 | XP_016882486.1 | 5 | 1154 | |||
XM_017026998.1 | XP_016882487.1 | 5 | 1048 | |||
XM_017026999.1 | XP_016882488.1 | 5 | 1049 | |||
XM_017027000.1 | XP_016882489.1 | 5 | 1047 | |||
XM_017027001.1 | XP_016882490.1 | 4 | 875 | |||
XM_017027002.1 | XP_016882491.1 | 4 | 875 | |||
GenBank mRNA | AF289606.1 | - | 5 | 601 | ||
AK075149.1 | - | 5 | 875 | |||
BC008405.1 | - | 5 | 1119 | 4043484 | ||
BC063127.1 | - | 5 | 1128 | 4768860 | ||
M33665.1 | - | 3 | 401 | |||
M94891.1 | - | 5 | 1108 | |||
U18468.1 | - | 5 | 1027 | |||
U18469.1 | - | 4 | 770 | |||
X17097.1 | - | 5 | 1114 |
Gene Symbol: PSG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130014.1 | NP_001123486.1 | 4 | 874 | ||
NM_002781.3 | NP_002772.3 | 4 | 874 | |||
XM_017027003.1 | XP_016882492.1 | 4 | 872 | |||
GenBank mRNA | AA151017.1 | - | 2 | 142 | ||
BC012607.1 | - | 4 | 841 | 4043912 | ||
BG621332.1 | - | 2 | 295 | |||
L36607.1 | - | 3 | 510 | |||
M25384.1 | - | 4 | 842 | |||
M73713.1 | - | 4 | 828 |
Gene Symbol: PSG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206650.1 | NP_001193579.1 | 4 | 758 | ||
NM_001290042.1 | NP_001276971.1 | 5 | 1124 | |||
NM_002783.2 | NP_002774.2 | 5 | 1124 | |||
GenBank mRNA | AL546656.3 | - | 4 | 701 | ||
BC030979.1 | - | 5 | 1119 | |||
BC136401.1 | - | 4 | 678 |
Gene Symbol: PSG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130167.1 | NP_001123639.1 | 5 | 1120 | ||
NM_001130168.1 | NP_001123640.1 | 4 | 754 | |||
NM_182707.2 | NP_874366.1 | 5 | 1120 | |||
XM_011526986.1 | XP_011525288.1 | 4 | 854 | |||
XM_011526987.2 | XP_011525289.1 | 4 | 912 | |||
XM_011526988.2 | XP_011525290.1 | 5 | 1103 | |||
GenBank mRNA | AK092458.1 | - | 1 | 1571 | ||
AK300230.1 | - | 4 | 753 | |||
AL545004.3 | - | 4 | 738 | |||
BC137500.1 | - | 5 | 1120 | 9021500 | ||
BC142628.1 | - | 5 | 1108 | 40147510 | ||
CR749812.1 | - | 5 | 1119 | |||
M30629.1 | - | 3 | 605 |
Gene Symbol: PSG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301707.1 | NP_001288636.1 | 4 | 875 | ||
NM_001301708.1 | NP_001288637.1 | 4 | 875 | |||
NM_001301709.1 | NP_001288638.1 | 3 | 596 | |||
NM_002784.4 | NP_002775.3 | 5 | 1154 | |||
XM_005259075.3 | XP_005259132.1 | 4 | 837 | |||
XM_017027004.1 | XP_016882493.1 | 5 | 1116 | |||
XM_017027005.1 | XP_016882494.1 | 4 | 870 | |||
XM_017027006.1 | XP_016882495.1 | 3 | 591 | |||
XM_017027007.1 | XP_016882496.1 | 5 | 1116 | |||
GenBank mRNA | AK300187.1 | - | 6 | 1000 | ||
AK300229.1 | - | 4 | 840 | |||
AK313256.1 | - | 5 | 1121 | |||
BC005925.1 | - | 4 | 839 | 4044135 | ||
BC020759.1 | - | 5 | 1119 | 4731739 | ||
CR407605.1 | - | 5 | 1022 | |||
M34421.1 | - | 5 | 1100 | |||
M34481.1 | - | 5 | 1091 | |||
M38046.1 | - | 4 | 880 | |||
M58591.1 | - | 5 | 1093 | |||
M94890.1 | - | 4 | 810 | |||
U25987.1 | - | 3 | 474 | |||
X17610.1 | - | 5 | 1100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
