siRNA Details
Gene Symbol: CSAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724857.3 | XP_006724920.1 | 2 | 637 | ||
XM_017030023.1 | XP_016885512.1 | 2 | 586 | |||
GenBank mRNA | AF268421.1 | - | 4 | 419 |
Gene Symbol: CSAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129826.2 | NP_001123298.1 | 2 | 511 | ||
NM_001129828.2 | NP_001123300.1 | 2 | 460 | |||
NR_130638.1 | 5 | 453 | ||||
GenBank mRNA | AF268420.1 | - | 4 | 367 | ||
BC007228.1 | - | 5 | 453 | |||
BC119734.1 | - | 2 | 433 | 40119148 | ||
BC119735.1 | - | 2 | 432 | 40119150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
