siRNA Details
Gene Symbol: CCDC144A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014695.2 | NP_055510.1 | 11 | 2402 | ||
XM_017025429.1 | XP_016880918.1 | 11 | 2419 | |||
XM_017025430.1 | XP_016880919.1 | 11 | 2492 | |||
GenBank mRNA | AB011137.2 | - | 2 | 804 | ||
AK125971.1 | - | 7 | 777 | |||
BC133019.1 | - | 11 | 2402 | 40147041 | ||
BC144225.1 | - | 11 | 2402 | 9052745 |
Gene Symbol: CCDC144B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036647.1 | 11 | 2495 | |||
GenBank mRNA | BX647826.1 | - | 11 | 2486 |
Gene Symbol: CCDC144CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023380.1 | 11 | 2473 | |||
GenBank mRNA | DV080283.1 | - | 4 | 497 |
Gene Symbol: POTEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005356.2 | NP_001005356.1 | 10 | 1493 | ||
NR_027480.1 | 11 | 1542 | ||||
GenBank mRNA | AY462874.1 | - | 10 | 1493 | ||
AY465172.1 | - | 12 | 1542 | |||
CV341500.1 | - | 3 | 199 |
Gene Symbol: POTEH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136213.1 | NP_001129685.1 | 10 | 1604 | ||
GenBank mRNA | AY466021.1 | - | 11 | 1803 |
Gene Symbol: POTEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145442.1 | NP_001138914.1 | 10 | 1493 | ||
GenBank mRNA | BC063888.1 | - | 2 | 153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
