siRNA Details
Gene Symbol: POTEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005356.2 | NP_001005356.1 | 11 | 1599 | ||
NR_027480.1 | 12 | 1648 | ||||
GenBank mRNA | AY462874.1 | - | 11 | 1599 | ||
AY465171.1 | - | 13 | 1857 | |||
AY465172.1 | - | 13 | 1648 | |||
CV341500.1 | - | 4 | 305 |
Gene Symbol: POTEH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136213.1 | NP_001129685.1 | 11 | 1710 | ||
GenBank mRNA | AY466021.1 | - | 12 | 1909 |
Gene Symbol: POTEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145442.1 | NP_001138914.1 | 11 | 1599 | ||
GenBank mRNA | BC063888.1 | - | 3 | 259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
