siRNA Details
Gene Symbol: POTEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005356.2 | NP_001005356.1 | 11 | 1699 | ||
NR_027480.1 | 12 | 1748 | ||||
GenBank mRNA | AY462874.1 | - | 11 | 1699 | ||
AY465171.1 | - | 13 | 1957 | |||
AY465172.1 | - | 13 | 1748 | |||
CV341500.1 | - | 4 | 405 |
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 15 | 2070 | ||
XM_011511216.2 | XP_011509518.1 | 11 | 1794 | |||
XM_017004159.1 | XP_016859648.1 | 11 | 1825 | |||
XM_017004160.1 | XP_016859649.1 | 7 | 793 | |||
XM_017004161.1 | XP_016859650.1 | 17 | 2507 | |||
GenBank mRNA | AY462868.1 | - | 12 | 1909 | ||
AY462871.1 | - | 15 | 2070 | |||
AY462872.1 | - | 14 | 2028 | |||
AY462873.1 | - | 13 | 1954 |
Gene Symbol: POTEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099771.2 | NP_001093241.1 | 17 | 2418 | ||
XM_017004833.1 | XP_016860322.1 | 15 | 2193 | |||
XM_017004834.1 | XP_016860323.1 | 10 | 915 | |||
XM_017004835.1 | XP_016860324.1 | 8 | 852 | |||
GenBank mRNA | EF523384.1 | - | 17 | 2420 |
Gene Symbol: POTEH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136213.1 | NP_001129685.1 | 11 | 1810 | ||
GenBank mRNA | AY466021.1 | - | 12 | 2009 |
Gene Symbol: POTEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145442.1 | NP_001138914.1 | 11 | 1699 | ||
GenBank mRNA | BC063888.1 | - | 3 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
