siRNA Details
Gene Symbol: POTEI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277406.1 | NP_001264335.1 | 15 | 2065 | ||
XM_011511670.2 | XP_011509972.1 | 15 | 2237 | |||
XM_017004732.1 | XP_016860221.1 | 16 | 2216 | |||
XM_017004733.1 | XP_016860222.1 | 10 | 962 | |||
XM_017004734.1 | XP_016860223.1 | 8 | 899 |
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 15 | 2117 | ||
XM_011511216.2 | XP_011509518.1 | 11 | 1841 | |||
XM_017004159.1 | XP_016859648.1 | 11 | 1872 | |||
XM_017004160.1 | XP_016859649.1 | 7 | 840 | |||
XM_017004161.1 | XP_016859650.1 | 17 | 2554 | |||
GenBank mRNA | AY462868.1 | - | 12 | 1956 | ||
AY462871.1 | - | 15 | 2117 | |||
AY462872.1 | - | 14 | 2075 | |||
AY462873.1 | - | 13 | 2001 |
Gene Symbol: POTEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005356.2 | NP_001005356.1 | 11 | 1746 | ||
NR_027480.1 | 12 | 1795 | ||||
GenBank mRNA | AY462874.1 | - | 11 | 1746 | ||
AY465172.1 | - | 13 | 1795 | |||
CV341500.1 | - | 4 | 452 |
Gene Symbol: POTEH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136213.1 | NP_001129685.1 | 11 | 1857 |
Gene Symbol: POTEJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277083.1 | NP_001264012.1 | 15 | 1954 | ||
XM_011511683.2 | XP_011509985.1 | 15 | 2126 | |||
XM_017004741.1 | XP_016860230.1 | 16 | 2099 | |||
XM_017004742.1 | XP_016860231.1 | 10 | 962 | |||
XM_017004743.1 | XP_016860232.1 | 8 | 894 |
Gene Symbol: POTEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145442.1 | NP_001138914.1 | 11 | 1746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
