siRNA Details
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102747.1 | 1 | 287 | |||
GenBank mRNA | AK309746.1 | - | 1 | 264 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 1 | 291 | |||
NR_033936.3 | 1 | 291 | ||||
GenBank mRNA | AK026448.1 | - | 1 | 76 | ||
AK309732.1 | - | 1 | 242 | |||
AK311170.1 | - | 1 | 201 | |||
BC007811.2 | - | 1 | 215 | |||
BC065260.1 | - | 1 | 208 | |||
BC072390.1 | - | 1 | 244 | |||
DB070142.1 | - | 1 | 291 |
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 1 | 274 | |||
GenBank mRNA | AK074778.1 | - | 1 | 242 | ||
AK307326.1 | - | 1 | 242 | |||
AK311265.1 | - | 1 | 196 | |||
BC054509.1 | - | 1 | 128 | |||
BC071951.1 | - | 1 | 231 | |||
BC096759.1 | - | 1 | 231 | |||
BU931675.1 | - | 253 |
Gene Symbol: LOC101929479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022812.1 | XP_016878301.1 | 1 | 264 |
Gene Symbol: LOC102724135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522305.2 | XP_011520607.1 | 1 | 1218 |
Gene Symbol: LOC642423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308544.1 | - | 1 | 237 |
Gene Symbol: UBE2Q2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004847.3 | 1 | 196 | |||
GenBank mRNA | BI916968.1 | - | 1 | 215 | ||
CN343524.1 | - | 1 | 184 | |||
DA194105.1 | - | 1 | 190 | |||
DA778756.1 | - | 1 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
