siRNA Details
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 1 | 244 | |||
GenBank mRNA | AK074778.1 | - | 1 | 212 | ||
AK307326.1 | - | 1 | 212 | |||
AK309056.1 | - | 1 | 166 | |||
AK311265.1 | - | 1 | 166 | |||
BC054509.1 | - | 1 | 98 | |||
BC071951.1 | - | 1 | 201 | |||
BC096759.1 | - | 1 | 201 | |||
BU931675.1 | - | 223 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 1 | 261 | |||
NR_033936.3 | 1 | 261 | ||||
NR_103496.2 | 1 | 261 | ||||
GenBank mRNA | AK026448.1 | - | 1 | 46 | ||
AK309420.1 | - | 1 | 212 | |||
AK309732.1 | - | 1 | 212 | |||
AK310769.1 | - | 1 | 214 | |||
AK311124.1 | - | 1 | 166 | |||
AK311170.1 | - | 1 | 171 | |||
BC007811.2 | - | 1 | 185 | |||
BC065260.1 | - | 1 | 178 | |||
BC072390.1 | - | 1 | 214 | |||
DB070142.1 | - | 1 | 261 |
Gene Symbol: LOC101929479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022812.1 | XP_016878301.1 | 1 | 234 |
Gene Symbol: LOC102724135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522304.2 | XP_011520606.1 | 1 | 1216 | ||
XM_011522305.2 | XP_011520607.1 | 1 | 1188 |
Gene Symbol: LOC642423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308544.1 | - | 1 | 207 |
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102747.1 | 1 | 257 | |||
NR_102748.1 | 1 | 257 | ||||
GenBank mRNA | AK291684.1 | - | 1 | 212 | ||
AK307285.1 | - | 1 | 202 | |||
AK309746.1 | - | 1 | 234 |
Gene Symbol: UBE2Q2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004847.3 | 1 | 166 | |||
GenBank mRNA | BI916968.1 | - | 1 | 185 | ||
CN343524.1 | - | 1 | 154 | |||
DA194105.1 | - | 1 | 160 | |||
DA527368.1 | - | 1 | 234 | |||
DA778756.1 | - | 1 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
