siRNA Details
Gene Symbol: HIST1H4J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021968.3 | NP_068803.1 | 1 | 247 | ||
GenBank mRNA | AF086564.1 | - | 1 | 264 | ||
BC017361.2 | - | 1 | 266 | 4647914 | ||
BC148271.1 | - | 1 | 269 | 8860214 |
Gene Symbol: HIST1H4I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003495.2 | NP_003486.1 | 1 | 247 | ||
GenBank mRNA | BC016336.1 | - | 1 | 278 | ||
BC066250.1 | - | 1 | 247 | 7002105 | ||
BC067496.1 | - | 1 | 247 | 7002103 | ||
BC067497.1 | - | 1 | 247 | 7002106 | ||
BC075806.1 | - | 1 | 259 |
Gene Symbol: HIST1H4K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003541.2 | NP_003532.1 | 1 | 247 | ||
GenBank mRNA | AK312028.1 | - | 1 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
