siRNA Details
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 7 | 1088 | ||
XM_017023622.1 | XP_016879111.1 | 8 | 1359 | |||
XM_017023623.1 | XP_016879112.1 | 9 | 1296 | |||
XM_017023624.1 | XP_016879113.1 | 8 | 1212 | |||
XM_017023625.1 | XP_016879114.1 | 8 | 1192 | |||
XM_017023626.1 | XP_016879115.1 | 9 | 1831 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 7 | 1041 | ||
XM_011523496.2 | XP_011521798.1 | 8 | 1097 | |||
XM_011523498.2 | XP_011521800.1 | 8 | 1131 | |||
XM_011523501.2 | XP_011521803.1 | 4 | 923 | |||
XM_017023248.1 | XP_016878737.1 | 8 | 1534 | |||
XM_017023249.1 | XP_016878738.1 | 7 | 1351 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 7 | 1063 | ||
XM_005255741.3 | XP_005255798.1 | 8 | 1875 | |||
XM_005255743.3 | XP_005255800.1 | 4 | 2454 | |||
XM_011545929.2 | XP_011544231.1 | 9 | 1695 | |||
XM_011545931.2 | XP_011544233.1 | 8 | 1328 | |||
XM_017023628.1 | XP_016879117.1 | 8 | 1078 | |||
XM_017023629.1 | XP_016879118.1 | 8 | 1621 | |||
XM_017023630.1 | XP_016879119.1 | 9 | 1348 | |||
XM_017023631.1 | XP_016879120.1 | 7 | 1692 | |||
XM_017023632.1 | XP_016879121.1 | 8 | 1078 | |||
GenBank mRNA | BC144439.1 | - | 7 | 1024 | ||
BC157836.1 | - | 7 | 1776 | 9057055 | ||
KF175222.1 | - | 7 | 1088 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 7 | 1126 | ||
NM_001287251.1 | NP_001274180.1 | 8 | 1212 | |||
XM_003118719.7 | XP_003118767.2 | 8 | 1687 | |||
XM_011545690.2 | XP_011543992.1 | 10 | 1848 | |||
XM_011545693.2 | XP_011543995.1 | 9 | 1775 | |||
XM_017022814.1 | XP_016878303.1 | 9 | 1923 | |||
XM_017022815.1 | XP_016878304.1 | 9 | 1443 | |||
XM_017022816.1 | XP_016878305.1 | 10 | 1860 | |||
XM_017022817.1 | XP_016878306.1 | 8 | 1835 | |||
XM_017022818.1 | XP_016878307.1 | 8 | 3160 | |||
XM_017022819.1 | XP_016878308.1 | 9 | 1850 | |||
XM_017022820.1 | XP_016878309.1 | 9 | 1567 | |||
XM_017022821.1 | XP_016878310.1 | 8 | 2433 | |||
XM_017022822.1 | XP_016878311.1 | 9 | 3436 | |||
XM_017022823.1 | XP_016878312.1 | 7 | 3072 | |||
XM_017022824.1 | XP_016878313.1 | 9 | 1702 | |||
XM_017022825.1 | XP_016878314.1 | 9 | 1420 | |||
XM_017022826.1 | XP_016878315.1 | 8 | 1384 | |||
XM_017022827.1 | XP_016878316.1 | 4 | 1131 | |||
GenBank mRNA | BC171825.1 | - | 8 | 1862 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003610.1 | 26 | 3966 | |||
GenBank mRNA | AL833251.1 | - | 26 | 3966 | ||
BC063633.1 | - | 24 | 2453 | |||
BX647358.1 | - | 2 | 5689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
