siRNA Details
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 7 | 894 | ||
XM_017023622.1 | XP_016879111.1 | 8 | 1165 | |||
XM_017023623.1 | XP_016879112.1 | 8 | 1165 | |||
XM_017023624.1 | XP_016879113.1 | 8 | 1018 | |||
XM_017023625.1 | XP_016879114.1 | 8 | 998 | |||
XM_017023626.1 | XP_016879115.1 | 9 | 1637 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 7 | 862 | ||
XM_011523496.2 | XP_011521798.1 | 8 | 918 | |||
XM_011523498.2 | XP_011521800.1 | 8 | 952 | |||
XM_011523501.2 | XP_011521803.1 | 4 | 744 | |||
XM_017023248.1 | XP_016878737.1 | 8 | 1355 | |||
XM_017023249.1 | XP_016878738.1 | 7 | 1172 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 7 | 932 | ||
XM_005255741.3 | XP_005255798.1 | 8 | 1744 | |||
XM_005255743.3 | XP_005255800.1 | 4 | 2323 | |||
XM_011545929.2 | XP_011544231.1 | 9 | 1564 | |||
XM_011545931.2 | XP_011544233.1 | 8 | 1197 | |||
XM_017023628.1 | XP_016879117.1 | 8 | 947 | |||
XM_017023629.1 | XP_016879118.1 | 8 | 1490 | |||
XM_017023630.1 | XP_016879119.1 | 9 | 1217 | |||
XM_017023631.1 | XP_016879120.1 | 7 | 1561 | |||
XM_017023632.1 | XP_016879121.1 | 8 | 947 | |||
GenBank mRNA | BC144439.1 | - | 7 | 893 | ||
BC157836.1 | - | 7 | 1582 | 9057055 | ||
KF175222.1 | - | 7 | 894 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 7 | 932 | ||
NM_001287251.1 | NP_001274180.1 | 8 | 1018 | |||
XM_003118719.7 | XP_003118767.2 | 8 | 1493 | |||
XM_011545690.2 | XP_011543992.1 | 10 | 1654 | |||
XM_011545693.2 | XP_011543995.1 | 9 | 1581 | |||
XM_017022814.1 | XP_016878303.1 | 9 | 1729 | |||
XM_017022815.1 | XP_016878304.1 | 9 | 1249 | |||
XM_017022816.1 | XP_016878305.1 | 9 | 1729 | |||
XM_017022817.1 | XP_016878306.1 | 8 | 1641 | |||
XM_017022818.1 | XP_016878307.1 | 8 | 2966 | |||
XM_017022819.1 | XP_016878308.1 | 9 | 1656 | |||
XM_017022820.1 | XP_016878309.1 | 9 | 1373 | |||
XM_017022821.1 | XP_016878310.1 | 8 | 2239 | |||
XM_017022822.1 | XP_016878311.1 | 9 | 3242 | |||
XM_017022823.1 | XP_016878312.1 | 7 | 2878 | |||
XM_017022824.1 | XP_016878313.1 | 9 | 1508 | |||
XM_017022825.1 | XP_016878314.1 | 9 | 1226 | |||
XM_017022826.1 | XP_016878315.1 | 8 | 1190 | |||
XM_017022827.1 | XP_016878316.1 | 4 | 937 | |||
GenBank mRNA | BC171825.1 | - | 8 | 1668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
