siRNA Details
Gene Symbol: SUMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042469.1 | NP_001035934.2 | 8 | 1831 | ||
NM_001042470.1 | NP_001035935.2 | 6 | 1624 | |||
NM_001130069.2 | NP_001123541.1 | 8 | 1731 | |||
NM_001146333.1 | NP_001139805.1 | 8 | 1719 | |||
NM_015411.2 | NP_056226.2 | 9 | 1876 | |||
XM_017011939.1 | XP_016867428.1 | 10 | 1994 | |||
XM_017011940.1 | XP_016867429.1 | 9 | 2166 | |||
GenBank mRNA | AF075046.1 | - | 1 | 327 | ||
AK075477.1 | - | 9 | 1835 | |||
AL050037.1 | - | 7 | 1597 | |||
AY359103.1 | - | 9 | 1808 | |||
BC000224.1 | - | 8 | 1611 | |||
BC006159.1 | - | 9 | 1773 | |||
BC015600.2 | - | 8 | 1627 | |||
BC065222.1 | - | 6 | 1515 | |||
BC084539.1 | - | 9 | 1807 | 6599080 |
Gene Symbol: AIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286588.1 | NP_001273517.1 | 4 | 2407 | ||
NM_001286589.1 | NP_001273518.1 | 4 | 2166 | |||
GenBank mRNA | BX640703.1 | - | 4 | 2108 |
Gene Symbol: C20orf196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303477.1 | NP_001290406.1 | 2 | 137 | ||
GenBank mRNA | BU656896.1 | - | 2 | 90 |
Gene Symbol: CDK5RAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124412.1 | - | 1 | 1281 |
Gene Symbol: CSPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127108.1 | - | 16 | 3210 |
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028911.1 | - | 7 | 989 |
Gene Symbol: DTWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308081.1 | NP_001295010.1 | 6 | 3999 | ||
NM_173666.3 | NP_775937.1 | 6 | 4220 | |||
XM_011543338.2 | XP_011541640.2 | 7 | 5613 | |||
XM_011543340.2 | XP_011541642.1 | 7 | 4112 | |||
GenBank mRNA | AK124328.1 | - | 1 | 1538 | ||
BX648369.1 | - | 6 | 3997 |
Gene Symbol: FAM138A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026818.1 | 2 | 407 | |||
GenBank mRNA | AY341950.1 | - | 2 | 403 |
Gene Symbol: FAM138B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026821.1 | 2 | 407 | |||
GenBank mRNA | AF346307.1 | - | 2 | 408 | ||
AY341948.1 | - | 2 | 403 |
Gene Symbol: FAM138C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026822.1 | 2 | 407 | |||
GenBank mRNA | AY341949.1 | - | 2 | 403 | ||
AY341953.1 | - | 2 | 403 |
Gene Symbol: FAM138D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026823.2 | 2 | 405 | |||
GenBank mRNA | AY341951.1 | - | 2 | 403 |
Gene Symbol: FAM138E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026819.1 | 2 | 407 |
Gene Symbol: FAM138F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026820.1 | 2 | 407 | |||
GenBank mRNA | AY341952.1 | - | 2 | 403 | ||
BC137207.1 | - | 2 | 239 |
Gene Symbol: GEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130009.2 | NP_001123481.2 | 14 | 4278 | ||
NM_182625.4 | NP_872431.4 | 14 | 4202 | |||
XM_005262613.4 | XP_005262670.1 | 14 | 4288 | |||
XM_006712005.3 | XP_006712068.1 | 14 | 4185 | |||
XM_011532820.2 | XP_011531122.1 | 14 | 4205 | |||
XM_011532821.2 | XP_011531123.1 | 14 | 4295 | |||
XM_011532822.2 | XP_011531124.1 | 14 | 4175 | |||
GenBank mRNA | AK025489.1 | - | 1 | 1721 | ||
CR936783.1 | - | 14 | 4288 |
Gene Symbol: GOLGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM842318.1 | - | 2 | 491 |
Gene Symbol: GTF3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005264272.4 | XP_005264329.1 | 2 | 307 |
Gene Symbol: IKBKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091090.1 | - | 1 | 60 | ||
AK093537.1 | - | 1 | 381 | |||
AK095687.1 | - | 1 | 1079 |
Gene Symbol: LOC100129550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024618.1 | 1 | 1306 | |||
GenBank mRNA | AK127315.1 | - | 1 | 880 |
Gene Symbol: LOC100130899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039988.1 | 1 | 3929 |
Gene Symbol: LOC105373221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091229.1 | - | 2 | 169 |
Gene Symbol: LRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122887.1 | - | 1 | 1344 |
Gene Symbol: MYLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR749364.1 | - | 1 | 1209 |
Gene Symbol: NDUFA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA189271.1 | - | 4 | 553 |
Gene Symbol: PIGO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251619.3 | XP_005251676.1 | 11 | 4743 | ||
XM_017015222.1 | XP_016870711.1 | 12 | 5235 |
Gene Symbol: SERPINB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004155.5 | NP_004146.1 | 7 | 3782 | ||
XM_005249184.4 | XP_005249241.1 | 7 | 3945 | |||
XM_011514678.2 | XP_011512980.1 | 7 | 3793 | |||
XM_017010942.1 | XP_016866431.1 | 8 | 3932 | |||
XM_017010943.1 | XP_016866432.1 | 8 | 4090 |
Gene Symbol: TNK2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094115.1 | - | 1 | 1233 |
Gene Symbol: ZNF655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009960.1 | NP_001009960.1 | 3 | 2585 | ||
NM_001083956.1 | NP_001077425.1 | 4 | 2863 | |||
NM_001085368.1 | NP_001078837.1 | 4 | 2690 | |||
NM_138494.2 | NP_612503.1 | 3 | 2758 | |||
XM_017012598.1 | XP_016868087.1 | 6 | 3763 | |||
XM_017012599.1 | XP_016868088.1 | 6 | 2921 | |||
XM_017012600.1 | XP_016868089.1 | 6 | 2934 | |||
XM_017012601.1 | XP_016868090.1 | 6 | 2920 | |||
XM_017012602.1 | XP_016868091.1 | 5 | 3658 | |||
XM_017012603.1 | XP_016868092.1 | 4 | 3783 | |||
XM_017012604.1 | XP_016868093.1 | 4 | 2700 | |||
XM_017012605.1 | XP_016868094.1 | 5 | 2711 | |||
XM_017012606.1 | XP_016868095.1 | 3 | 2616 | |||
XM_017012607.1 | XP_016868096.1 | 3 | 2602 | |||
XM_017012608.1 | XP_016868097.1 | 6 | 3796 | |||
XM_017012609.1 | XP_016868098.1 | 5 | 2810 | |||
XM_017012610.1 | XP_016868099.1 | 5 | 2757 | |||
GenBank mRNA | AK054750.1 | - | 1 | 73 | ||
AK098231.1 | - | 1 | 464 | |||
AK122929.1 | - | 1 | 1842 | |||
AK128057.1 | - | 2 | 3822 | |||
BC024770.1 | - | 1 | 2104 | |||
BC037407.1 | - | 3 | 2548 | 5752559 | ||
CR749270.1 | - | 3 | 2366 |
Gene Symbol: ZSCAN20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271171.3 | XP_005271228.1 | 8 | 7631 | ||
XM_006710874.3 | XP_006710937.1 | 8 | 7634 | |||
XM_006710876.3 | XP_006710939.1 | 6 | 7472 | |||
XM_017002237.1 | XP_016857726.1 | 8 | 7677 | |||
XM_017002238.1 | XP_016857727.1 | 8 | 7633 | |||
XM_017002239.1 | XP_016857728.1 | 7 | 7617 | |||
XM_017002240.1 | XP_016857729.1 | 7 | 7549 | |||
XM_017002241.1 | XP_016857730.1 | 6 | 8120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
