siRNA Details
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018660.2 | NP_061130.1 | 10 | 3292 | ||
GenBank mRNA | AK026076.1 | - | 1 | 1159 | ||
AL137588.1 | - | 1 | 608 | |||
BC002919.1 | - | 1 | 844 | |||
BC008870.2 | - | 1 | 558 | |||
BC063660.1 | - | 1 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
