siRNA Details
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291420.1 | NP_001278349.1 | 8 | 1291 | ||
NM_198181.3 | NP_937824.3 | 9 | 1423 | |||
XM_011521601.2 | XP_011519903.1 | 8 | 1974 | |||
XM_017022226.1 | XP_016877715.1 | 7 | 1410 | |||
GenBank mRNA | DR157204.1 | - | 1 | 167 |
Gene Symbol: GOLGA6L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267536.2 | NP_001254465.2 | 9 | 1811 | ||
XM_017022481.1 | XP_016877970.1 | 10 | 1655 | |||
XM_017022482.1 | XP_016877971.1 | 10 | 1655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
