siRNA Details
Gene Symbol: GOLGA6L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267536.2 | NP_001254465.2 | 9 | 2078 | ||
XM_017022481.1 | XP_016877970.1 | 10 | 1922 | |||
XM_017022482.1 | XP_016877971.1 | 10 | 1922 | |||
GenBank mRNA | AK090778.1 | - | 1 | 183 |
Gene Symbol: GOLGA6L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164465.3 | NP_001157937.2 | 9 | 1962 | ||
GenBank mRNA | CR936602.1 | - | 12 | 2613 |
Gene Symbol: GOLGA6L5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003246.4 | 10 | 2037 | |||
GenBank mRNA | BX647899.1 | - | 1 | 434 |
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128268.1 | - | 5 | 1261 | ||
CR936812.1 | - | 6 | 4117 | |||
DR157204.1 | - | 1 | 458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
