siRNA Details
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286835.1 | NP_001273764.1 | 9 | 831 | ||
XM_005272748.1 | XP_005272805.1 | 8 | 703 | |||
XM_005272749.1 | XP_005272806.1 | 8 | 703 | |||
XM_005272750.1 | XP_005272807.1 | 7 | 643 | |||
XM_006716914.1 | XP_006716977.1 | 8 | 703 | |||
XM_017014456.1 | XP_016869945.1 | 9 | 634 | |||
XM_017014458.1 | XP_016869947.1 | 8 | 703 | |||
XM_017014459.1 | XP_016869948.1 | 10 | 882 | |||
GenBank mRNA | AK097639.1 | - | 7 | 1019 | ||
AK292585.1 | - | 9 | 771 | |||
BC067803.1 | - | 9 | 769 | 5294979 | ||
BC141856.1 | - | 9 | 769 |
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291821.1 | NP_001278750.1 | 7 | 627 | ||
NM_201453.3 | NP_958861.2 | 8 | 687 | |||
XM_005277637.2 | XP_005277694.1 | 8 | 766 | |||
XM_006717116.1 | XP_006717179.1 | 8 | 766 | |||
XM_011518706.2 | XP_011517008.1 | 3 | 217 | |||
XM_017014731.1 | XP_016870220.1 | 8 | 3138 | |||
XM_017014733.1 | XP_016870222.1 | 3 | 155 | |||
GenBank mRNA | AK297912.1 | - | 7 | 589 | ||
AY344633.1 | - | 8 | 663 |
Gene Symbol: CBWD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085457.1 | NP_001078926.1 | 8 | 706 | ||
XM_017015019.1 | XP_016870508.1 | 7 | 706 | |||
XM_017015020.1 | XP_016870509.1 | 7 | 706 | |||
XM_017015021.1 | XP_016870510.1 | 7 | 706 | |||
XM_017015023.1 | XP_016870512.1 | 7 | 706 | |||
XM_017015026.1 | XP_016870515.1 | 7 | 706 | |||
XM_017015027.1 | XP_016870516.1 | 7 | 706 | |||
XM_017015028.1 | XP_016870517.1 | 8 | 574 | |||
XM_017015029.1 | XP_016870518.1 | 4 | 525 | |||
XM_017015030.1 | XP_016870519.1 | 7 | 706 | |||
XM_017015031.1 | XP_016870520.1 | 3 | 217 | |||
XM_017015032.1 | XP_016870521.1 | 7 | 706 | |||
XM_017015034.1 | XP_016870523.1 | 2 | 80 | |||
XM_017015035.1 | XP_016870524.1 | 8 | 766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
