siRNA Details
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 8 | 802 | ||
XM_005263600.2 | XP_005263657.1 | 8 | 802 | |||
XM_005263603.2 | XP_005263660.1 | 9 | 836 | |||
XM_005263605.3 | XP_005263662.1 | 9 | 837 | |||
XM_011510667.1 | XP_011508969.1 | 8 | 802 | |||
XM_011510668.2 | XP_011508970.1 | 5 | 362 | |||
XM_011510669.2 | XP_011508971.1 | 9 | 846 | |||
XM_017003407.1 | XP_016858896.1 | 8 | 802 | |||
XM_017003408.1 | XP_016858897.1 | 10 | 966 | |||
GenBank mRNA | AF452722.1 | - | 8 | 658 | ||
AK225251.1 | - | 9 | 816 | |||
AK290072.1 | - | 8 | 802 | |||
AK303653.1 | - | 9 | 841 | |||
AY343910.1 | - | 8 | 687 | |||
BC120991.1 | - | 8 | 673 | 40116409 | ||
BC120992.1 | - | 8 | 673 | 40116411 |
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291821.1 | NP_001278750.1 | 7 | 651 | ||
NM_201453.3 | NP_958861.2 | 8 | 711 | |||
XM_005277637.2 | XP_005277694.1 | 8 | 790 | |||
XM_006717116.1 | XP_006717179.1 | 8 | 790 | |||
XM_011518706.2 | XP_011517008.1 | 3 | 241 | |||
XM_017014731.1 | XP_016870220.1 | 8 | 3162 | |||
XM_017014733.1 | XP_016870222.1 | 3 | 179 | |||
GenBank mRNA | AK297912.1 | - | 7 | 613 | ||
AY344633.1 | - | 8 | 687 |
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286835.1 | NP_001273764.1 | 9 | 855 | ||
XM_005272748.1 | XP_005272805.1 | 8 | 727 | |||
XM_005272749.1 | XP_005272806.1 | 8 | 727 | |||
XM_005272750.1 | XP_005272807.1 | 7 | 667 | |||
XM_006716914.1 | XP_006716977.1 | 8 | 727 | |||
XM_017014456.1 | XP_016869945.1 | 9 | 658 | |||
XM_017014458.1 | XP_016869947.1 | 8 | 727 | |||
XM_017014459.1 | XP_016869948.1 | 10 | 906 | |||
GenBank mRNA | AK292585.1 | - | 9 | 795 | ||
BC067803.1 | - | 9 | 793 | 5294979 |
Gene Symbol: CBWD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF293368.1 | - | 1 | 35 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
