siRNA Details
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024916.3 | NP_001020087.2 | 10, 11 | 894 | ||
NM_001286835.1 | NP_001273764.1 | 14, 15 | 2146 | |||
XM_005272748.1 | XP_005272805.1 | 12, 13 | 998 | |||
XM_005272749.1 | XP_005272806.1 | 11, 12 | 941 | |||
XM_005272750.1 | XP_005272807.1 | 11, 12 | 938 | |||
XM_005272751.1 | XP_005272808.1 | 11, 12 | 911 | |||
XM_006716914.1 | XP_006716977.1 | 12, 13 | 998 | |||
XM_017014455.1 | XP_016869944.1 | 11, 12 | 911 | |||
XM_017014456.1 | XP_016869945.1 | 13, 14 | 929 | |||
XM_017014457.1 | XP_016869946.1 | 12, 13 | 842 | |||
XM_017014459.1 | XP_016869948.1 | 14, 15 | 1177 | |||
XM_017014461.1 | XP_016869950.1 | 13, 14 | 1090 | |||
GenBank mRNA | AK097639.1 | - | 12, 13 | 1531 | ||
AK292585.1 | - | 13, 14 | 1066 | |||
BC043420.1 | - | 2, 3 | 796 | |||
BC067803.1 | - | 14, 15 | 2084 | 5294979 | ||
BC082271.1 | - | 10, 11 | 788 | |||
BC141856.1 | - | 13, 14 | 1064 | |||
BM711778.1 | - | 3, 4 | 156 |
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 13, 14 | 1099 | ||
NM_001145356.1 | NP_001138828.1 | 11, 12 | 944 | |||
NM_018491.3 | NP_060961.3 | 12, 13 | 1001 | |||
XM_005251513.4 | XP_005251570.1 | 11, 12 | 949 | |||
XM_005251514.1 | XP_005251571.1 | 11, 12 | 914 | |||
XM_005251515.1 | XP_005251572.1 | 10, 11 | 857 | |||
XM_011517961.2 | XP_011516263.1 | 12, 13 | 1036 | |||
XM_011517962.2 | XP_011516264.1 | 12, 13 | 1001 | |||
XM_011517963.1 | XP_011516265.1 | 9, 10 | 539 | |||
GenBank mRNA | AF212253.1 | - | 11, 12 | 912 | ||
AF257330.2 | - | 12, 13 | 959 | |||
AK290648.1 | - | 13, 14 | 1071 | |||
AK297292.1 | - | 12, 13 | 983 | |||
AK315855.1 | - | 12, 13 | 925 | |||
AY343911.1 | - | 12, 13 | 958 | |||
BC005996.1 | - | 12, 13 | 1001 | 4069589 | ||
BC009573.1 | - | 13, 14 | 1052 | 4133536 | ||
BC013432.1 | - | 12, 13 | 972 | 4133722 | ||
BC132759.1 | - | 12, 13 | 935 | 40146781 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 12, 13 | 1073 | ||
XM_005263600.2 | XP_005263657.1 | 11, 12 | 1016 | |||
XM_005263602.2 | XP_005263659.1 | 11, 12 | 986 | |||
XM_005263603.2 | XP_005263660.1 | 13, 14 | 1107 | |||
XM_005263604.2 | XP_005263661.1 | 10, 11 | 929 | |||
XM_005263605.3 | XP_005263662.1 | 12, 13 | 1051 | |||
XM_011510667.1 | XP_011508969.1 | 12, 13 | 1073 | |||
XM_011510668.2 | XP_011508970.1 | 9, 10 | 633 | |||
XM_011510669.2 | XP_011508971.1 | 13, 14 | 1117 | |||
XM_017003408.1 | XP_016858897.1 | 14, 15 | 1237 | |||
GenBank mRNA | AF452722.1 | - | 12, 13 | 929 | ||
AK124867.1 | - | 3, 4 | 2638 | |||
AK225251.1 | - | 13, 14 | 1087 | |||
AK290072.1 | - | 12, 13 | 1073 | |||
AY343910.1 | - | 12, 13 | 958 | |||
BC120991.1 | - | 12, 13 | 944 | 40116409 | ||
BC120992.1 | - | 12, 13 | 944 | 40116411 |
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY344633.1 | - | 12, 13 | 958 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146097, BC053745, BC027643, AK076027, BC018472, and others.
Gene Ontology Categories:
