siRNA Details
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291821.1 | NP_001278750.1 | 14 | 1242 | ||
NM_201453.3 | NP_958861.2 | 15 | 1302 | |||
XM_005277637.2 | XP_005277694.1 | 14 | 1324 | |||
XM_005277639.2 | XP_005277696.1 | 14 | 1294 | |||
XM_005277640.2 | XP_005277697.1 | 13 | 1237 | |||
XM_006717116.1 | XP_006717179.1 | 14 | 1251 | |||
XM_011518706.2 | XP_011517008.1 | 10 | 832 | |||
XM_017014730.1 | XP_016870219.1 | 13 | 1234 | |||
XM_017014731.1 | XP_016870220.1 | 15 | 3753 | |||
XM_017014732.1 | XP_016870221.1 | 9 | 745 | |||
XM_017014733.1 | XP_016870222.1 | 10 | 6052 | |||
GenBank mRNA | AK297912.1 | - | 14 | 1204 | ||
AY344633.1 | - | 15 | 1278 |
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 16 | 1419 | ||
NM_001145356.1 | NP_001138828.1 | 14 | 1264 | |||
NM_018491.3 | NP_060961.3 | 15 | 1321 | |||
XM_005251513.4 | XP_005251570.1 | 14 | 1269 | |||
XM_005251514.1 | XP_005251571.1 | 14 | 1234 | |||
XM_005251515.1 | XP_005251572.1 | 13 | 1177 | |||
XM_011517961.2 | XP_011516263.1 | 15 | 1356 | |||
XM_011517963.1 | XP_011516265.1 | 12 | 859 | |||
GenBank mRNA | AF212253.1 | - | 14 | 1232 | ||
AF257330.2 | - | 15 | 1279 | |||
AK290648.1 | - | 16 | 1391 | |||
AK315855.1 | - | 15 | 1245 | |||
AY343911.1 | - | 15 | 1278 | |||
BC005996.1 | - | 15 | 1321 | 4069589 | ||
BC009573.1 | - | 16 | 1372 | 4133536 | ||
BC013432.1 | - | 15 | 1292 | 4133722 | ||
BC132759.1 | - | 15 | 1255 | 40146781 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 15 | 1393 | ||
XM_005263600.2 | XP_005263657.1 | 14 | 1336 | |||
XM_005263602.2 | XP_005263659.1 | 14 | 1306 | |||
XM_005263603.2 | XP_005263660.1 | 16 | 1427 | |||
XM_005263604.2 | XP_005263661.1 | 13 | 1249 | |||
XM_011510667.1 | XP_011508969.1 | 14 | 1263 | |||
XM_011510668.2 | XP_011508970.1 | 12 | 953 | |||
XM_011510669.2 | XP_011508971.1 | 16 | 1437 | |||
XM_017003408.1 | XP_016858897.1 | 17 | 1557 | |||
GenBank mRNA | AF452722.1 | - | 15 | 1249 | ||
AK124867.1 | - | 6 | 2958 | |||
AK225251.1 | - | 16 | 1407 | |||
AK290072.1 | - | 15 | 1393 | |||
AY343910.1 | - | 15 | 1278 | |||
BC120991.1 | - | 15 | 1264 | 40116409 | ||
BC120992.1 | - | 15 | 1264 | 40116411 | ||
DA296279.1 | - | 1 | 131 |
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024916.3 | NP_001020087.2 | 13 | 1214 | ||
NM_001286835.1 | NP_001273764.1 | 17 | 2466 | |||
XM_005272748.1 | XP_005272805.1 | 15 | 1318 | |||
XM_005272749.1 | XP_005272806.1 | 14 | 1261 | |||
XM_005272750.1 | XP_005272807.1 | 14 | 1258 | |||
XM_005272751.1 | XP_005272808.1 | 14 | 1231 | |||
XM_006716914.1 | XP_006716977.1 | 14 | 1188 | |||
XM_017014455.1 | XP_016869944.1 | 13 | 1101 | |||
XM_017014456.1 | XP_016869945.1 | 16 | 1249 | |||
XM_017014457.1 | XP_016869946.1 | 15 | 1162 | |||
XM_017014459.1 | XP_016869948.1 | 17 | 1497 | |||
XM_017014461.1 | XP_016869950.1 | 16 | 1410 | |||
GenBank mRNA | AK097639.1 | - | 15 | 1851 | ||
BC043420.1 | - | 3 | 2626 | |||
BC067803.1 | - | 17 | 2404 | 5294979 | ||
BC082271.1 | - | 13 | 1108 | |||
BC141856.1 | - | 16 | 1384 |
Gene Symbol: CBWD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085457.1 | NP_001078926.1 | 15 | 1321 | ||
XM_017015019.1 | XP_016870508.1 | 15 | 1366 | |||
XM_017015020.1 | XP_016870509.1 | 14 | 1321 | |||
XM_017015021.1 | XP_016870510.1 | 14 | 1309 | |||
XM_017015022.1 | XP_016870511.1 | 14 | 1279 | |||
XM_017015023.1 | XP_016870512.1 | 13 | 1264 | |||
XM_017015024.1 | XP_016870513.1 | 13 | 1234 | |||
XM_017015025.1 | XP_016870514.1 | 13 | 1222 | |||
XM_017015026.1 | XP_016870515.1 | 14 | 1236 | |||
XM_017015028.1 | XP_016870517.1 | 16 | 1234 | |||
XM_017015029.1 | XP_016870518.1 | 12 | 1185 | |||
XM_017015031.1 | XP_016870520.1 | 11 | 877 | |||
XM_017015033.1 | XP_016870522.1 | 10 | 790 | |||
XM_017015034.1 | XP_016870523.1 | 10 | 740 | |||
XM_017015035.1 | XP_016870524.1 | 14 | 1324 | |||
XM_017015036.1 | XP_016870525.1 | 13 | 1237 | |||
GenBank mRNA | AF293368.1 | - | 8 | 626 | ||
AK293173.1 | - | 15 | 1370 | |||
BC069006.1 | - | 16 | 1462 | |||
CR627025.1 | - | 2 | 3057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
