siRNA Details
Gene Symbol: LOC101929567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110257.1 | 6 | 6745 | |||
GenBank mRNA | BX641130.1 | - | 6 | 6752 |
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020297.3 | NP_064693.2 | 38 | 7199 | ||
XM_005253284.3 | XP_005253341.1 | 40 | 7985 | |||
XM_005253286.3 | XP_005253343.1 | 41 | 8045 | |||
XM_005253287.4 | XP_005253344.1 | 41 | 8161 | |||
XM_005253288.3 | XP_005253345.1 | 39 | 7869 | |||
XM_005253289.3 | XP_005253346.1 | 39 | 7946 | |||
XM_005253290.3 | XP_005253347.1 | 38 | 7844 | |||
XM_006719025.3 | XP_006719088.1 | 40 | 8122 | |||
XM_011520545.2 | XP_011518847.1 | 40 | 8041 | |||
GenBank mRNA | BX640893.1 | - | 1 | 2134 |
Gene Symbol: FSIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092099.1 | - | 8 | 2655 |
Gene Symbol: GC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298433.1 | - | 12 | 1465 |
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 3091 |
Gene Symbol: WDPCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX538331.1 | - | 14 | 2608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
