siRNA Details
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 355 | ||
AF336884.1 | - | 1 | 424 | |||
AF338194.1 | - | 1 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
