siRNA Details
Gene Symbol: ODF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007022.2 | NP_001007023.2 | 18 | 2832 | ||
NM_020729.2 | NP_065780.2 | 18 | 2791 | |||
XM_011541819.2 | XP_011540121.1 | 18 | 2837 | |||
XM_011541821.2 | XP_011540123.1 | 17 | 2678 | |||
XM_017001879.1 | XP_016857368.1 | 17 | 2750 | |||
GenBank mRNA | AB033055.1 | - | 18 | 2752 | ||
BC047368.1 | - | 18 | 2730 | 5276959 |
Gene Symbol: LOC105372751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135520.1 | 6 | 1070 | |||
GenBank mRNA | AK124194.1 | - | 6 | 1070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
