siRNA Details
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 3068 | |||
GenBank mRNA | BC036758.2 | - | 7 | 3080 | 5563071 |
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020297.3 | NP_064693.2 | 38 | 7176 | ||
XM_005253284.3 | XP_005253341.1 | 40 | 7962 | |||
XM_005253286.3 | XP_005253343.1 | 41 | 8022 | |||
XM_005253287.4 | XP_005253344.1 | 41 | 8138 | |||
XM_005253288.3 | XP_005253345.1 | 39 | 7846 | |||
XM_005253289.3 | XP_005253346.1 | 39 | 7923 | |||
XM_005253290.3 | XP_005253347.1 | 38 | 7821 | |||
XM_006719025.3 | XP_006719088.1 | 40 | 8099 | |||
XM_011520545.2 | XP_011518847.1 | 40 | 8018 | |||
GenBank mRNA | BX640893.1 | - | 1 | 2111 |
Gene Symbol: ATP9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC053561.1 | - | 5 | 1545 |
Gene Symbol: CFAP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC041426.1 | - | 7 | 1869 |
Gene Symbol: FSIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092099.1 | - | 8 | 2632 |
Gene Symbol: LOC100129434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125368.1 | 3 | 2850 | |||
GenBank mRNA | AK096196.1 | - | 3 | 3059 |
Gene Symbol: LOC101926898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125837.1 | 3 | 1326 | |||
GenBank mRNA | BC042616.1 | - | 3 | 1328 |
Gene Symbol: LOC101929567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110257.1 | 6 | 6722 | |||
GenBank mRNA | BX641130.1 | - | 6 | 6729 |
Gene Symbol: LOC283728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095617.1 | - | 2 | 3468 |
Gene Symbol: UGDH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047679.1 | 5 | 2951 | |||
GenBank mRNA | AK056558.1 | - | 5 | 2771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
