siRNA Details
Gene Symbol: AFDN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027906.1 | 2 | 1926 | |||
GenBank mRNA | AB016898.1 | - | 2 | 1839 | ||
AK091690.1 | - | 3 | 1828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
