siRNA Details
Gene Symbol: TP53TG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015381.1 | 1 | 176 | |||
GenBank mRNA | AB007455.1 | - | 1 | 199 | ||
AB007456.1 | - | 1 | 199 | |||
AB007457.1 | - | 1 | 170 | |||
AB007458.1 | - | 1 | 170 | |||
AK292782.1 | - | 1 | 193 | |||
BC002709.2 | - | 1 | 176 | |||
BC061927.1 | - | 1 | 129 | |||
BC068535.1 | - | 1 | 210 | |||
BC103631.1 | - | 1 | 145 | |||
BT009845.1 | - | 1 | 62 | |||
CR542141.1 | - | 1 | 62 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
