siRNA Details
Gene Symbol: ZG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152338.3 | NP_689551.2 | 4 | 579 | ||
XM_011545921.2 | XP_011544223.1 | 4 | 658 | |||
GenBank mRNA | AK125559.1 | - | 4 | 537 | ||
BC029149.1 | - | 3 | 511 | 5187790 | ||
BC150656.1 | - | 4 | 550 | 9057027 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
