siRNA Details
Gene Symbol: ZG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152338.3 | NP_689551.2 | 4 | 1456 | ||
GenBank mRNA | AK125559.1 | - | 4 | 1414 | ||
BC150656.1 | - | 4 | 1429 | 9057027 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
