siRNA Details
Gene Symbol: CTAG1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001327.2 | NP_001318.1 | 1 | 256 | ||
GenBank mRNA | AF038567.1 | - | 1 | 291 | ||
AJ003149.1 | - | 1 | 253 | |||
AJ275978.1 | - | 1 | 125 | |||
BC130362.1 | - | 1 | 260 | 40146393 | ||
BC130364.1 | - | 1 | 260 | 40146395 | ||
U87459.1 | - | 1 | 256 |
Gene Symbol: CTAG1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139250.1 | NP_640343.1 | 1 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
