siRNA Details
Gene Symbol: ERVK-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF543113.1 | - | 1 | 86 |
Gene Symbol: ERVK-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF153348.1 | - | 1 | 86 |
Gene Symbol: ERVK-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF153341.1 | - | 1 | 85 |
Gene Symbol: ERVK-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF543114.1 | - | 1 | 85 |
Gene Symbol: ERVK-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF153351.1 | - | 1 | 86 |
Gene Symbol: ERVK-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ069911.1 | - | 1 | 746 | ||
DQ069912.1 | - | 1 | 746 | |||
DQ069913.1 | - | 1 | 749 | |||
DQ069914.1 | - | 1 | 746 | |||
DQ069915.1 | - | 1 | 746 | |||
DQ069916.1 | - | 1 | 746 | |||
DQ069917.1 | - | 1 | 746 | |||
DQ069918.1 | - | 1 | 746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
