siRNA Details
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 14 | 2147 | ||
NM_031921.5 | NP_114127.3 | 16 | 2030 | |||
XM_011542243.2 | XP_011540545.1 | 15 | 1766 | |||
XM_011542244.1 | XP_011540546.1 | 16 | 2197 | |||
GenBank mRNA | AB033099.1 | - | 14 | 2236 | ||
AK024028.1 | - | 14 | 1615 | |||
AK127857.1 | - | 10 | 2801 | |||
AK290586.1 | - | 16 | 2022 | |||
AL834179.1 | - | 5 | 2936 | |||
BC002542.2 | - | 16 | 1956 | 3138578 | ||
BC018701.1 | - | 16 | 1956 | 3139886 |
Gene Symbol: ATAD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025865.1 | - | 9 | 1026 | ||
AK056099.1 | - | 16 | 1856 | |||
AK091144.1 | - | 16 | 2138 | |||
AK092833.1 | - | 1 | 2142 | |||
BC007803.2 | - | 16 | 1987 | 4136170 | ||
BC011814.2 | - | 16 | 1992 | 3162989 | ||
BC014101.2 | - | 16 | 2016 | 3503438 | ||
BC033109.1 | - | 16 | 1990 | 4877449 | ||
BC063607.1 | - | 16 | 1984 | 4939339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
