siRNA Details
Gene Symbol: ATAD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170535.1 | NP_001164006.1 | 11 | 1246 | ||
NM_001170536.1 | NP_001164007.1 | 11 | 1076 | |||
NM_018188.3 | NP_060658.3 | 11 | 1390 | |||
XM_017001628.1 | XP_016857117.1 | 11 | 1246 | |||
GenBank mRNA | AK001571.1 | - | 11 | 1229 | ||
AK025865.1 | - | 4 | 246 | |||
AK056099.1 | - | 11 | 1076 | |||
AK091144.1 | - | 11 | 1358 | |||
AK310572.1 | - | 1268 | ||||
BC007803.2 | - | 11 | 1207 | 4136170 | ||
BC011814.2 | - | 11 | 1212 | 3162989 | ||
BC014101.2 | - | 11 | 1236 | 3503438 | ||
BC033109.1 | - | 11 | 1210 | 4877449 | ||
BC063607.1 | - | 11 | 1204 | 4939339 | ||
GU189416.1 | - | 11 | 1150 |
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 9 | 1367 | ||
NM_031921.5 | NP_114127.3 | 11 | 1250 | |||
XM_005244806.3 | XP_005244863.1 | 11 | 1273 | |||
XM_011542241.2 | XP_011540543.1 | 11 | 1273 | |||
XM_011542243.2 | XP_011540545.1 | 10 | 986 | |||
XM_011542244.1 | XP_011540546.1 | 11 | 1417 | |||
GenBank mRNA | AB033099.1 | - | 9 | 1456 | ||
AF343078.1 | - | 11 | 1183 | |||
AK024028.1 | - | 9 | 835 | |||
AK127857.1 | - | 4 | 1901 | |||
AK290586.1 | - | 11 | 1242 | |||
AL834179.1 | - | 3 | 1088 | |||
BC002542.2 | - | 11 | 1176 | 3138578 | ||
BC018701.1 | - | 11 | 1176 | 3139886 |
Gene Symbol: ATAD3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039211.2 | NP_001034300.2 | 7 | 1590 | ||
GenBank mRNA | AK091918.1 | - | 7 | 1591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
